Silencer® Select Negative Control No. 2 siRNA

Catalog number: 4390847

Invitrogen™  Related applications: RNAi | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Silencer® SelectNegative Control #2 siRNA is extensively validated and an ideal control for many aspects of an siRNA experiment. 40 nmol is provided.

• Premium-quality siRNA, highly purified and ready to use
• Functionally proven to have minimal effects on cell proliferation and viability
• Include Silencer® Select modifications for enhanced specificity
• For use in human, mouse, and rat cells

What are Silencer® Select siRNAs?
Silencer® Select siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications, to yield siRNAs with unrivalled efficacy, potency, and specificity. The result is fewer failed experiments due to poor silencing, and cleaner, more consistent phenotypic data.

Silencer® Select Negative Control #2 siRNA:
Negative control siRNAs—siRNAs with sequences that do not target any gene product—are essential for determining the effects of siRNA delivery and for providing a baseline to compare siRNA-treated samples. Silencer® Select Negative Control #2 siRNA has been carefully designed and extensively tested, and has no significant sequence similarity to mouse, rat, or human gene sequences. This negative control siRNA has been tested by microarray analysis and shown to have minimal effects on gene expression. It has also been tested in multi-parametric, cell-based assays and shown to have no significant effect on cell proliferation, viability, or morphology in the cell lines tested.

Quality Control:
Silencer® Select siRNAs are manufactured in a state-of-the-art facility under the highest quality standards. Rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. Each siRNA is also assessed by gel electrophoresis to confirm that the strands anneal properly.

Accessory Product: siPORT™ NeoFX™ Transfection Agent (SKU #AM4510 and #AM4511)
siPORT™ NeoFX™ Transfection Agent is a versatile, lipid-based transfection agent for the efficient transfection of siRNA into cultured cells. The protocol can be adapted to a wide range of cells and experimental designs, including high-throughput applications. Silencer® Select siRNA controls have specific chemical modifications; they are designed to be used with Silencer® Select siRNAs. If you are using other siRNA products, such as BLOCK-iT™ siRNAs, Ambion® Silencer® siRNAs, or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Type: Negative Control
Label or Dye: Unconjugated
Product Line: Silencer™, Silencer™ Select, Ambion™
Product Size: 40 nmol
RNAi Type: siRNA
Shipping Condition: Room Temperature

Contents & storage

The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.

Documents

Manuals & protocols