Conditioning Reagent, 3500 Series

Catalog number: 4393718

Applied Biosystems™  Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 3500 series conditioning reagent is available as a ready-to-use pouch, and is used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down. The pouch has adequate volume for a one-time use. Incorporated into the label is a radio frequency identification (RFID) tag. The top part of the pouch fitment is sealed with a plastic film, which should be removed prior to installation on to the instrument.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer
Form: Liquid
Product Size: 1 unit
Reagent Type: Conditioning Reagent for Sequencing Instrument
Shipping Condition: Wet Ice

Contents & storage

Store pouches at 2 to 8°C.

Documents

Manuals & protocols