Applied Biosystems™

Computer RAM Upgrade Kit, laptop D510/D520

Catalog number:  4399679

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This upgrade kit is intended for use with the 7300⁄7500⁄7500 Fast Sequence Detection Systems.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 7300 System, 7500 Fast System, 7500 System
Product Size: 1 kit
Shipping Condition: Room Temperature
Type: Computer Accessories

Contents & storage

Store at room temperature.

Documents

Manuals & protocols