Hi-Di™ Formamide (CE-IVD)

Catalog number: 4404307

Applied Biosystems™  Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hi-Di™ Formamide is a highly de-ionized formamide, formulated with a stabilizer, for use as an injection solvent on Applied Biosystems® 3500 Dx Series genetic analyzers. This highly deionized formamide is used to resuspend samples before electrokinetic injection in capillary electrophoresis systems and is appropriate for all applications. It is provided in a 5 mL bottle format to minimize freeze-thaw cycles.
For In Vitro Diagnostic Use.

Specifications

Chemical Name: Hi-Di Formamide
Form: Liquid
Product Line: High-Di™
Product Size: 5mL bottle
Reagent Type: Formamide
Shipping Condition: Dry Ice

Contents & storage

Store at -15 to -25°C.

Documents

Manuals & protocols