TaqMan® Protein Assays Core Reagents Base Kit contains the reagents required to perform the ligation and PCR amplification steps of TaqMan® protein expression experiments. The Universal PCR Assay contains optimized concentrations of TaqMan® probe and oligonucleotide primers (forward and reverse) that are designed to amplify a region of the protease-treated ligation product for detection on an Applied Biosystems real-time PCR system.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Product Line
TaqMan™
For Use With (Application)
Proximity Ligation Assay
Detection Method
Primer-Probe Detection
Label or Dye
FAM
Product Type
Reagents for Protein Expression
Quantity
100 Reactions
Sample Type
Cells
No. of Reactions
100 reactions
Primer Probe Compatibility
TaqMan™
PCR Method
qPCR Protein Quantification
Reaction Speed
Fast
Shipping Condition
Wet Ice
Target Specificity
Not Target-Specific
Contents & Storage
The TaqMan™ Protein Assays Core Reagents Base Kit contains: