ViiA™ 7 Array Card RNaseP Verification Kit

Catalog number: 4432265

Applied Biosystems™  Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ViiA™ 7 System TaqMan® Low Density Array Instrument Verification RNase P kit is used to validate the performance of the Applied Biosystems® ViiA™ 7 Real-Time PCR Systems with the TaqMan® Low Density Array Block Upgrade.

The ViiA™ 7 System TaqMan® Low Density Array Instrument Verification RNase P kit contains an empty TaqMan® Low Density Array and 8 vials of RNase P detection reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each vial contains premixed reagents (1X TaqMan® Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.

The ViiA™ 7 System TaqMan® Low Density Array Instrument Verification RNase P kit must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems® recommends that the performance of your instrument be verified every six months.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibration Type: Instrument Verification Plate, RNase P Calibration
For Use With (Equipment): ViiA™ 7 System
Format: 384-well microfluidic card
Product Line: Applied Biosystems™, ViiA™
Product Size: 1 kit

Contents & storage

Store this kit at -15 to -25°C.

Documents

Manuals & protocols