Applied Biosystems™

VIC™ Dye Calibration Plate, Fast 96-well

Catalog number:  4432607

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Fast 96-Well Dye Calibration Plate (VIC®) is used to maintain your real-time PCR instrument with Fast 96-well block. Dye calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.
For In Vitro Diagnostic Use.

Specifications

Calibration Type: Spectral Calibration
For Use With (Equipment): QuantStudio™ Dx, ViiA™ 7 Dx System
Label or Dye: VIC®
Product Size: 1 x 96 well plate
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products