Applied Biosystems™

Normalization Kit, Fast 96-well

Catalog number:  4443774

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Fast 96-Well Normalization Kit is used to maintain your real-time PCR instrument with Fast 96-well block. Normalization plates are used to calibrate different instruments. The kit contains one FAM™/ROX™ normalization plate and one VIC®/ROX™ normalization plate. Please see your instrument user guide for full instructions on use.
For In Vitro Diagnostic Use.

Specifications

Calibration Type: Spectral Calibration
For Use With (Equipment): QuantStudio™ Dx, ViiA™ 7 Dx System
Label or Dye: Proprietary Mix
Product Size: 2 x 96 well plates
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products