Applied Biosystems™

Megaplex™ RT Primers, Rodent Pool B v3.0 This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  4444292

 Related applications: Real Time PCR (qPCR) | miRNA & Non-Coding RNA Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Designed to streamline experimental workflows, Megaplex™ RT Primers provide a single reaction solution when preparing cDNA for real-time PCR analysis on a TaqMan® Array MicroRNA Card. Rodent Pool B v2.0 contains RT primers for 306 and 135 unique microRNAs for mouse and rat respectively plus 4 species specific controls. The microRNA targets represented in this pool tend be less well characterized, more narrowly expressed and⁄or expressed at a lower level when compared to Rodent Pool A. The pool can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Primer Type: RT Primers
Product Line: Megaplex™, TaqMan™
Product Size: 50 reactions
Species: Mouse, Rat
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store at -20°C

Documents

Manuals & protocols

Recommended products