Applied Biosystems™

Megaplex™ Primer Pools, Rodent Pools Set v3.0

Catalog number:  4444766

 Related applications: Real Time PCR (qPCR) | miRNA & Non-Coding RNA Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Megaplex™ Primer Pools, Rodent Pools Set v3.0 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Rodent Pools Sets. Megaplex™ RT Primers, Rodent Pools Set collectively contains RT primers for 641 and 373 unique microRNAs for mouse and rat respectively plus 4 species specific controls. For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The Megaplex™ Primer Pools, Rodent Pools Set is intended for use with the TaqMan® Rodent MicroRNA Array Set v3.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Primer Type: PreAmp Primers, RT Primers
Product Line: Megaplex™, TaqMan™
Product Size: 50 reactions
Species: Mouse, Rat

Contents & storage

Store at -20°C

Documents

Manuals & protocols