TaqMan™ Array Human MicroRNA B Cards v3.0

Catalog number:  4444910

Applied Biosystems™  Related applications: Real Time PCR (qPCR) | miRNA & Non-Coding RNA Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The TaqMan® Array Human MicroRNA B Card v3.0 contains 384 TaqMan® MicroRNA Assays enabling accurate quantitation of 377 human microRNAs in as little as 5 hours. Three TaqMan MicroRNA Assay endogenous controls are included to aid in data normalization and one TaqMan® MicroRNA Assay not related to human is included as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool B v3.0 is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool B v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.
For Research Use Only. Not for use in diagnostic procedures.


Form: Dried Down
Format: 384-well microfluidic card
Label or Dye: FAM™
Product Line: Megaplex™
Product Size: 4 pack
Reaction Speed: Standard
Research Category: Epigenetics, Gene Regulation
Species: Human

Contents & storage

Store at 4°C


Manuals & protocols