TaqMan® Protein Assays Oligo Probe Kit

Catalog number: 4448549

Applied Biosystems™  Related applications: Real Time PCR (qPCR) | Stem Cell Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

TaqMan® Protein Assays Oligo Probe Kit contains the 5’ and 3’ Prox-Oligos required to prepare the assay probes for TaqMan® Protein Assays experiments. The kit contains the oligonucleotide pair used in combination with the TaqMan® Protein Assays Buffer Kit (4448571)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Primer-Probe Detection
For Use With (Equipment): 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode
Label or Dye: FAM™
PCR Method: qPCR Protein Quantification
Primer-Probe Compatibility: TaqMan®
Product Line: TaqMan™
Product Size: 4,000 reactions
Target Specificity: Not Target-Specific
Technique: Proximity Ligation Assay
Shipping Condition: Wet Ice

Contents & storage

TaqMan® Protein Assays Oligo Probe Kit contains the oligonucleotide pair used in combination with the TaqMan® Protein Assays Buffer Kit (4448571)

Documents

Manuals & protocols