TaqMan™ Protein Assays Fast Master Mix

Catalog number: 4448616

Applied Biosystems™  Related applications: Real Time PCR (qPCR) | Stem Cell Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The enzyme concentration, ionic strength, and the pH of the TaqMan® Protein Assays Fast Master Mix are optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan® Protein Assays reagents. The DNA polymerase included in the master mix is designed for maximum PCR efficiency and specificity when used with the ligation chemistry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Primer-Probe Detection
For Use With (Equipment): 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode
Hot Start: No
Label or Dye: FAM™
PCR Method: qPCR Protein Quantification
Passive Reference Dye: ROX (Pre-mixed)
Polymerase: AmpliTaq® Fast DNA Polymerase
Primer-Probe Compatibility: TaqMan®
Product Line: TaqMan™
Product Size: 500 reactions
Reaction Speed: Fast
Sample Type (General): Cells, Tissue
Target Specificity: Not Target-Specific
Technique: Proximity Ligation Assay
Shipping Condition: Wet Ice

Contents & storage

TaqMan® Protein Assays Fast Master Mix is optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan® ProteinAssays reagents

Documents

Manuals & protocols