Applied Biosystems™
21 CFR Part 11 Software Module for ViiA™ 7 System
Catalog number:  4453723
 Related applications: Real Time PCR (qPCR)
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This software module supports compliance with 21CFR Part 11 FDA guidelines on electronic records security and prevents unauthorized access to your ViiA7 instruments. When loaded on the instrument, this software will:
• Authenticate each user’s log-in information
• Lock out access and requite re-authentication if user is inactive for a set period of time
• Log any unauthorized attempts to access ViiA7 System software in the Event Log
• Verify each user’s permission to perform predefined Controlled Activities.
• Permits E-signature function for data verification.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): ViiA™ 7 System
Software Type: Compliance
Operating System: Windows
Product Line: ViiA™
Product Size: 1 software
Software Category: Instrument Access and Security
Contents & storage
One software CD.
Documents
Manuals & protocols