Applied Biosystems™
HRM Software Module for ViiA™ 7 System
Catalog number:  4453724
 Related applications: Genotyping & Genomic Profiling | Real Time PCR (qPCR)
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Expands the functionality of the ViiA7 system, allowing High Resolution Melting (HRM) analysis to be performed on PCR amplicons. HRM analysis can discriminate genetic variation by amplicon length, GC content, sequence variability or strand complementarity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): ViiA™ 7 System
Software Type: Genotyping, Primer Design
Operating System: Windows
Product Line: ViiA™
Product Size: 1 software
Software Category: Analysis
Contents & storage
One software CD
Documents
Manuals & protocols
Recommended products