Applied Biosystems™
SOLiD™ SAGE™ Module D with Barcoding Adaptor
Catalog number:  4456595
 Related applications: RNA Sequencing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The SOLiD™ SAGE™ Module D with Barcoding Adaptor allows legacy SOLiD™ SAGE™ kits to interface with SOLiD™ RNA Barcoding Kits. In order to barcode SOLiD™ SAGE™ samples the SOLiD™ RNA Barcoding Kit is also recommended to purchase separately to use in conjunction with the SOLiD™ SAGE™ Module D. This part is only recommended for SOLiD™ users with current SOLiD™ SAGE™ library reagents who are interested in barcoding. The SOLiD™ SAGE™ update also offers an updated free data analysis package available from the SOLiD™ software user community site which provides greater mapping flexibility and higher data mapping.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): SOLiD™
Product Line: SOLiD™
Product Size: 16 reactions
Contents & storage
SOLiD™ SAGE™ Module D store at -20°C includes; Barcode Adaptor A, Wash Buffer D, Platinum PCR Supermix
Documents
Manuals & protocols