3500xL Dx Genetic Analyzer CS2 This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  4461447

Applied Biosystems™  Related applications: DNA Sequencing | Diagnostic Testing | Fragment Analysis | Genotyping & Genomic Profiling | RNA Sequencing | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 3500 Dx Genetic Analyzer CS2 is the latest addition to our family of Applied Biosystems® genetic analyzers based on capillary electrophoresis. The 3500 Dx Genetic Analyzer CS2 is an IVD-labeled instrument, making it the first Sanger sequencing platform cleared for sequencing analysis of DNA.

Confidence in Your Results
• The 3500 Dx Genetic Analyzer CS2 is an IVD-labeled platform for Sequencing-Based Typing using SeCore® HLA typing kits. Sanger sequencing is known as the "gold-standard" for its accuracy, reliability, and ease of use. It is also used for confirmatory sequencing by clinical labs.
• High accuracy: 99% concordance as shown in clinical trials using SeCore® HLA typing kits
• Proven through decades of results—including sequencing of the first human genome—Applied Biosystems® genetic analysis systems are trusted standards for Sanger sequencing.

You can now feel even more comfortable adopting the 3500 Dx Genetic Analyzer CS2 platform for the development of assays for FDA submission.

Simplicity and Efficiency
• Reduced instrument hands-on time with simplified installation and removal of consumables and accessories
• Controlled and efficient run setup with intuitive user interface, preconfigured plate templates, and software-controlled functions (plate setup, data collection, analysis)
• Instrument run-times as short as 1 hour; sample-to-result in as little as 1.5 days
• Maintenance calendar with programmable prompts and service history
• Security and electronic signature, RFID (radio frequency identification) tracking of consumables, built-in quality controls, and real-time data quality evaluation

The 3500 Dx Genetic Analyzer CS2 is a partner in your laboratory, making it easy for you to comply with various industry standards and simplifying the training of new users.

Flexibility
• Collect data in either IVD (diagnostic for HLA typing) or RUO mode
• Develop other applications in RUO mode, including de novo sequencing and re-sequencing, as well as fragment analysis applications such as microsatellite analysis, MLPA®, LOH, MLST, and SNP confirmation or screening
• Partner with us to develop assays for FDA submission
• Different configurations to meet your throughput needs: 24-capillary (3500xL Dx) and 8-capillary (3500 Dx) versions

The 3500 Dx Genetic Analyzer CS2 is a trusted capillary electrophoresis-based sequencing platform for development of your new assays. Capillary electrophoresis-based applications are supported by a large number of publications over the many years that these applications have been used successfully in laboratory testing.

IVD-labeled Accessories and Consumables
The purchase of a 3500 Dx Genetic Analyzer CS2 includes a computer system and associated Data Collection software, as well as an installation kit containing the initial set of accessories and reagents needed to start using the instrument right away. The 3500 Dx Genetic Analyzer CS2 installation includes a full install qualification and operational qualification, as well as instrument training to introduce you to your new genetic analyzer.

The 3500 Dx is backed by a comprehensive Service & Support program with options to purchase additional services depending upon your laboratory maintenance and audit requirements.

Intended Use
The Applied Biosystems® 3500 Dx and 3500xL Dx Genetic Analyzers CS2 with 3500 Dx Series Software 2011 (v1) are in vitro diagnostic devices intended for the sequencing (detection and identification) of fluorescently labeled DNA by capillary electrophoresis.

The Applied Biosystems® 3500 Dx/3500xL Dx Genetic Analyzers CS2 with 3500 Dx Series Software 2011 (v1) are indicated for use with FDA-cleared or approved sequencing assays specifying their use and only by technologists trained in laboratory techniques, procedures, and use of the analyzer.

Note: The 3500 Dx Genetic Analyzer CS2 data collection software uses Windows 7 SP0 operating system. This makes the currently available RUO secondary analysis software incompatible for use on the same system. A separate computer is required to do secondary analysis if operating in RUO mode.
Available in select countries only.

Specifications

Analysis Software: uTYPE® Dx SBT Analysis Software
Capillary Length: 50 cm
Compatible Polymers: Dx: POP-6, RUO: POP-4/6/7
Dimensions: 61 cm W (122 cm W open door) x 61 cm D x 72 cm H
External Computer: OptiPlex™ XE Duo 3 GHz Processor
For Use With (Equipment): 3500xL Dx Genetic Analyzer
Format: 96-well plate
Green Features: Energy efficient, Fewer resources used, Less waste
Laser Features: Long-life single-line 505 nm solid-state laser
Memory: 4 GB Ram, 500 GB Hard Drive
Number of Capillaries: 24 Capillaries
Operating Environment: 15°C-30°C (<2° fluctuation during run)
Operating System: Windows 7
Optics: CCD Camera
Polymer Consumption: 8 samples/run (384 or 960 sample pouches)
Product Size: 1 system
Read Length: ≥ 450 bp (RapidSeq50_POP6), ≥ 600 bp (FastSeq0_POP6)
Throughput: ≥ 368 samples/day (FastSeq50_POP6), ≥ 504 samples/day (RapidSeq50_POP6)
Weight: 82 kg (approximately)

Contents & storage

This package consists of the 24 capillary instrument, Data Collection Software, a Dell Workstation and monitor as well as reagent kits for system qualification. Store at room temperature.

Documents

Manuals & protocols

Recommended products