Applied Biosystems™

SOLiD™ Run Cycle Buffer Kit, Double

Catalog number:  4461840

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ Buffers are used for ligation sequencing on the 5500⁄5500xl Genetic Analyzers. The SOLiD™ Run Cycle Buffer Kit (Double) contains a set of six buffers required during all types of sequencing runs for ligation sequencing in the forward and reverse directions. The kit consists of SOLiD™ Buffers A, B, C, E, F, and G.

SOLiD™ Buffers A, B, and E are provided in two parts, and components must be mixed before use. SOLiD™ Buffer F is provided as a 10X solution and requires dilution to 1X before pouring into its reagent tank. SOLiD™ Buffer G is poured into its reagent tank before use.

The kit contains sufficient reagents to run 950 to 1632 cycles, or up to 136 cycles per lane.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: SOLiD™
Product Size: 1 kit
Recommended Applications: De Novo Sequencing, Shotgun Sequencing

Contents & storage

This kit contains buffers for two 5500⁄5500xl FlowChips.

Store SOLiD™ Buffers A and B at 2°C to 8°C until ready to mix and use. Store the SOLiD™ Buffer E kit at -25°C to -15°C until ready to mix and use. Store SOLiD™ Buffers C, F, and G at room temperature until ready to use.

Documents

Manuals & protocols