SOLiD™ REV SR Kit, Double

Catalog number: 4463030

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ Sequencing Reagents (SR) Kits contain enzymes and probes needed for ligation sequencing with 2-base encoded (2BE) probes on the 5500⁄5500xl Genetic Analyzer. The SOLiD™ REV SR Kit (Double) is used for paired-end sequencing runs for ligation sequencing in the reverse direction. The kit contains sufficient ligase, reverse phosphatase, and reverse 2BE probes to run 280 to 420 cycles, or up to 35 cycles per lane. In addition to the SOLiD™ Run Cycle Buffers and SOLiD™ Buffer D (Reset), this kit requires the use of the SOLiD™ REV Buffer.

Double-sized buffers have sufficient reagents for covering two FlowChips.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Recommended Applications: De Novo Sequencing, Shotgun Sequencing
Form: Liquid
Product Line: SOLiD™
Product Size: 1 kit
Shipping Condition: Dry Ice

Contents & storage

This kit contains probes and enzymes for two 5500⁄5500xl FlowChips.

Store the SOLiD™ REV SR Kit at -25°C to -15°C until ready for use.

Documents

Manuals & protocols