Applied Biosystems™

PrepFiler™ Forensic DNA Extraction Kit, updated

Catalog number: 4463351
Applied Biosystems™

PrepFiler™ Forensic DNA Extraction Kit, updated

Catalog number: 4463351
Catalog Number
4463351
Unit Size
1 kit
Price (USD)
Price: 720.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44633511 kit
Price: 720.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PrepFiler Forensic DNA Extraction Kit uses magnetic particles with an optimized multi-component surface chemistry to deliver superior DNA yield from most forensic sample types, including body fluids, and stains and swabs of body fluids. Whether it’s an automated or manual application, this kit is designed to help improve the overall yield and concentration of DNA recovered from both routine and challenging samples, while enabling removal of PCR inhibitors. This means greater quantities of highly purified DNA for improved downstream genotyping results. This kit is intended for manual use with 1.5 mL tubes. For automated workflows, please see the PrepFiler Automated Forensic DNA Extraction Kit.

• Designed to improve the overall yield, concentration, and purity of DNA isolated from both routine and challenging forensic samples
• Contains specially developed magnetic particles and reagents that optimize DNA binding and elution of DNA in a small volume
• Offers streamlined protocols to process an extensive variety of forensic samples
• Part of an integrated solution including novel reagents, consumables, and automated options to provide reliable, simplified, and time-saving DNA purification
• Improves downstream genotyping (STR analysis) success rate, to help ensure that critical and often limited casework samples can be successfully analyzed

Improved Packaging and Configuration
Several packaging and configuration improvements have been implemented for the updated PrepFiler kits:
• An additional wash buffer, PrepFiler Wash Buffer B, has now been incorporated into the kits to minimize the potential for detergent carryover from PrepFiler Wash Buffer A, which can inhibit downstream PCR applications when the maximum volume of DNA extract is used for subsequent PCR amplification.
• The kit packaging has been updated to include all reagents, including magnetic particles, in the same kit box, with a single expiration date.
• Shipping will occur at ambient temperature, and the kits can also be stored upon receipt at ambient temperature.
• In this kit configuration, plastic consumables are not included (PrepFiler Filter Columns for substrate removal and PrepFiler Spin Tubes for sample processing in tube format). For the kit with plastics, see Cat. No. 4463350.

Specifications

Format
Filter Column, Tube
Isolation Technology
Magnetic Bead
Sample Type
Forensic Sample
For Use With (Application)
Forensic Testing
High-throughput Compatibility
Not High-throughput Compatible (Manual)
No. of Reactions
100
Product Line
PrepFiler
Shipping Condition
Room Temperature
Target
Forensic DNA

Contents & Storage

The kit includes all reagents, including magnetic particles, in the same kit box, with a single expiration date.

PrepFiler Lysis Buffer
PrepFiler Magnetic Particles
PrepFiler Wash Buffer A Concentrate
PrepFiler Wash Buffer B Concentrate
PrepFiler Elution Buffer

Note: In this kit configuration, plastic consumables are not included (PrepFiler Filter Columns for substrate removal and PrepFiler Spin Tubes for sample processing in tube format). For the plastics, see Cat. No. A36853.

Store all kit components at room temperature (18—25 °C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text