Invitrogen™

Pyk2 Antibody Sampler Pack

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Proline-rich/Ca2+-activated tyrosine kinase (Pyk2) is a member of the FAK family of non-receptor, proline-rich protein tyrosine kinases. Pyk2 signaling is initiated by a variety of extracellular stimuli including integrin ligation, CD surface marker ligation (e.g., CD3, CD28, TCR, VCAM), bioactive peptides, growth factors, cytokines, chemokines, and certain stress stimuli (reactive oxygen species and Ca++ flux).

Format:
Unconjugated

Application:
WB, IHC

Clone:
Polyclonal

Storage:
FREEZE - 20°C

References:
Chandoke, S.K., et al. (2004) Beta 3 integrin phosphorylation is essential for Arp3 organization into leukocyte alpha v beta 3-vitronectin adhesion contacts. J. Cell Sci. 117(Pt 8): 1431-1441 (cites the use of cat. # 44-618G and 44-876G). Catalog number 44638G is same antibody as former catalog number 44638. 44638G contains 50% glycerol, which improves the stability and longevity of this antibody.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human, Rat
Validated Application: Western Blot
Host Species: Rabbit
Clonality: Polyclonal
Clone ID: Sampler Pack
Target Gene: PYK2
Gene ID: 2185
Label or Dye: Unconjugated
Target Modification State: Phospho
Purification: Purified
Product Size: 4 vials
Research Category: Signal Transduction
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products