Invitrogen™

mirVana™ miRNA Mimic, Negative Control #1

Catalog number:  4464058

 Related applications: miRNA & Non-Coding RNA Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

mirVana™ miRNA Mimic Negative Control #1 is designed for use as a negative control for experiments using mirVana™ miRNA Mimics. miRNA mimics are small, chemically modified, double-stranded RNA molecules designed to specifically bind to and mimic endogenous miRNA molecules and enable miRNA functional analysis by up-regulation of miRNA activity. We have now released the second generation of miRNA mimics, mirVana™ miRNA Mimics, which:

• Exhibit maximum and consistent effects in vitro at low concentration
• Are nontoxic
• Offer superior specificity due to unique Star-strand modification
• Target the most updated Sanger content
• Can be used in vitro and in vivo, offering you consistency throughout your entire research project

mirVana™ miRNA Mimic Negative Control #1
mirVana™ miRNA Mimic Negative Control #1 is a random sequence miRNA mimic molecule that has been extensively tested in human cell lines and tissues and validated to not produce identifiable effects on known miRNA function. Transfect the Negative Control using the same methodology as for your positive control (such as miR-1 Positive Control) and experimental mirVana™ miRNA Mimics. Use target gene expression from Negative Control-transfected samples as a baseline for evaluation of the effect of the control and experimental miRNA mimic on target gene expression.

First-generation mimic controls
Our first-generation miRNA mimics (and controls), Pre-miR™ miRNA Precursors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Mimics. These first-generation mimics are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Mimics and their corresponding controls.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Type: Negative Control
Label or Dye: Unconjugated
Product Line: mirVana™
Product Size: 5 nmol
Purification: Purified
RNAi Type: Pre-miRNA
Shipping Condition: Room Temperature

Contents & storage

Store at or below –20°C. Do not store in a frost-free freezer.

Documents

Manuals & protocols