E-Gel® Imager UV Light Base

Catalog number: 4466602

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-Gel® Imager system is your personal imaging system for documenting and analyzing agarose gels and E-Gel® cassettes. The E-Gel® Imager UV Light Base is one of three interchangeable bases that work with the E-Gel® Imager Camera Hood to comprise the E-Gel® Imager system. The E-Gel® Imager UV Light base is a compact UV light transilluminator for self poured gels as well as E-Gel® agarose gel cassettes stained with ethidium bromide. E-Gel® Imager UV Light Base has a built in safety mechanism that requires the E-Gel® Imager Camera Hood to be affixed over the top in order to be operational. Alternatively the E-Gel® Imager Gel Excision Kit will allow the base to transilluminate without the Camera Hood attached. E-Gel® Imager UV Light Base has an internal power supply port to allow operation of the E-Gel® iBase™ power system inside the system without running cables or wires under the hood or through a door.

• Powerful UV transillumination
• Sleek, compact design
• Choose the best configuration for any stain with interchangeable bases

Strong, Steady Ultraviolet Transillumination
The E-Gel® Imager UV Light Base is an excellent transilluminator specially designed to work as part of the E-Gel® Imager system. The UV Light Base provides intense uniform illumination across the entirety of the glass surface of the table. The internal light sources have been positioned to minimize hot and cold spots as well as edge effects and to minimize shadowing. The E-Gel® Imager UV Light Base produces very bright and intense UV light. Proper personal protective equipment should always be used when working with UV light.

Small Footprint Fits Most Benchtop
The E-gel® Imager UV Light Base has been designed as a part of the compact E-Gel® Imager system. The contoured design is sleek and modern, to standout on any bench without overwhelming even a modest amount of space. E-Gel® Imager is a departure from the heavy, cumbersome imaging systems of years past. As part of the E-Gel® Imager system the UV Light Base is small and light enough to move around the lab as needed, but attractive enough to want to store in plain sight.

Multiple Bases for Multiple Applications
Three interchangeable bases provide excellent flexibility to use multiple gel stains. The UV Light Base is best for ethidium bromide and the Blue Light Base can be used for many SYBR® dyes like SYBR® Safe DNA gel stain. The E-Gel® Adaptor Base has been designed to provide power to either the E-Gel® iBase™ power system coupled to the E-Gel® Safe Imager™, or the E-Gel® Go! Base with its integrated blue light transilluminator so E-Gel® cassettes can be run right inside the E-Gel® Imager. This allows for real time image capture while samples are running on the gel.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): E-Gel® Imager
Label or Dye: Ethidium bromide
Product Line: E-Gel®
Product Size: 1 each
System: E-Gel® Imager
Shipping Condition: Room Temperature

Contents & storage

1 x E-Gel® Imager UV Light Base.

Store at room temperature.

Documents

Manuals & protocols

Recommended products