E-Gel™ Imager Adaptor Base

Catalog number:  4466604

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-Gel® Imager system is your personal imaging system for documenting and analyzing agarose gels and E-Gel® cassettes. The E-Gel® Imager Adaptor Base is one of three interchangeable bases that work with the E-Gel® Imager Camera Hood to comprise the E-Gel® Imager system. The E-Gel® Imager Adaptor Base provides power to either the E-Gel® iBase™ power system coupled to the E-Gel® Safe Imager™ or E-Gel® Go! Base to allow E-Gel® cassettes to be run right inside the imaging system. With the E-Gel® Imager Adaptor Base for the first time you’ll be able to document electrophoresis as your samples run.

• Compatible with your favorite E-Gel® devices
• Sleek, compact design
• Choose the best configuration for any stain with interchangeable bases

Document Electrophoresis in Real-Time, for the First Time
The E-Gel® Imager Adaptor Base is one of the most innovative components of the new E-Gel® Imager system. The Adaptor Base has been specifically designed to provide power to either the E-Gel® iBase™/E-Gel® Safe Imager™ base combinations or to the new E-Gel® Go! Base without having to run wires through a door or under the edge of the hood. This allows outside light to be completely excluded from the E-Gel® Imager system while a gel cassette is run. E-Gel® Imager Adaptor Base is the first tool that will allow you to capture real time images of E-Gel® cassettes as your samples run.

Small Footprint Fits Any Benchtop
The E-gel® Imager Blue Light Base has been designed as a part of the compact E-Gel® Imager system. The contoured design is sleek and modern, to standout on any bench without overwhelming even a modest amount of space. E-Gel® Imager is a departure from the heavy, cumbersome imaging systems of years past. As part of the E-Gel® Imager system the Blue Light Base is small and light enough to move around the lab as needed, but attractive enough to want to store in plain sight.

Multiple Bases for Multiple Applications
Three interchangeable bases provide excellent flexibility to use multiple gel stains. The UV Light Base is best for ethidium bromide and the Blue Light Base can be used for many SYBR® dyes like SYBR® Safe. The E-Gel® Adaptor Base has been designed to provide power to either the E-Gel® iBase™ power system coupled to the E-Gel® Safe Imager™ real-time transilluminator, or the E-Gel® Go! Base with its integrated blue light transilluminator so E-Gel® cassettes can be run right inside the E-Gel® Imager. This allows real time image capture while samples are running on the gel.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Products: E-Gel® Go! Base, E-Gel® iBase™
For Use With (Equipment): E-Gel® Imager
Product Line: E-Gel®
Product Size: 1 each
System: E-Gel® Imager
Shipping Condition: Room Temperature

Contents & storage

1 x E-Gel® Imager Adaptor Base.

Store at room temperature.

Documents

Manuals & protocols

Recommended products