Related applications: Nucleic Acid Gel Electrophoresis & Blotting
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Filter Type: | Emission Filters |
---|---|
For Use With (Equipment): | E-Gel® Imager |
Label or Dye: | Ethidium bromide, SYBR® Safe |
Product Line: | E-Gel™ |
Product Size: | 1 each |
Type: | Optical Filters |
Shipping Condition: | Room Temperature |