Invitrogen™

E-Gel™ Imager Qdot™ 625 Filter

Catalog number:  4466607

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-Gel® Imager system is your personal imaging system for documenting and analyzing agarose gels and E-Gel® cassettes. E-Gel® Imager is a very powerful and flexible system that allows you to document gel images using multiple nucleic acid dyes and even Qdot® nanocrystals. Each E-Gel® Imager system includes a universal filter along with the camera hood. E-Gel® Imager Qdot® 625 Filter is optimized for applications utilizing Qodot® 625 nanocrystal technology.

• Change filters easily without needing any tools
• Choose the perfect filter for your specific application

Filter Trays Allow Easy Swapping with No Extra Tools
Every E-Gel® Imager Camera Hood comes with a pre-installed universal filter. Alternative filters can be easily utilized by simply removing the universal filter tray and sliding in the filter tray of your choosing into the camera hood.

The Right Filter for Your Application
For most applications the universal filter installed in the camera hood at the time of purchase will be acceptable. There are two specific alternative filters that are currently available. First, there’s the E-Gel® Imager UV/SYBR® Filter, which is the best choice when documenting gels stainded with a SYBR® stain using UV transillumination. Second, there’s the E-Gel® Imager Qdot® 625 filter that can be used with Qdot® nanocrystal with an emission peak of 625nm.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Filter Type: Emission Filters
For Use With (Equipment): E-Gel® Imager
Label or Dye: Qdot® 625
Product Line: E-Gel™
Product Size: 1 each
Type: Optical Filters
Shipping Condition: Room Temperature

Contents & storage

1 x E-Gel® Imager Qdot® 625 Filter.

Store at room temperature.

Documents

Manuals & protocols

Recommended products