Invitrogen™

E-Gel™ Imager Quantitation USB Dongle

Catalog number:  4466610

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-Gel® Imager Quantitation Software USB Dongle is your key to our Gel Quant Express software and comes included with each E-Gel® Imager system. To use this powerful quantitation and analysis software you must insert an activation dongle into a free USB port on your PC whenever the software is in use. To use the software on more than one computer simultaneously additional E-Gel® Imager Quantitation Software USB Dongles can be purchased.

• Access the powerful Gel Quant Express software using the USB dongle

The Key to Quantitation
Gel Quant Express is a powerful and intuitive software tool included with each E-Gel® Imager system. In order to utilize the software a HASP key in the form of a USB dongle must be inserted into a free USB port on your computer. Each E-Gel® Imager system includes a single Quantitiation Software USB Dongle that will allow a single instance of the Gel quant Express software to be used at a given time. A separate USB dongle is required for each instance of the Gel Quant Express Software that will be used at a given time.

A Compact Solution for Agarose Gel Imaging, Analysis, and Documentation
The E-Gel® Imager system is your personal imaging system for documenting and analyzing agarose gels and E-Gel® cassettes. E-Gel® Imager is a very powerful and flexible system that allows you to document gel images using multiple nucleic acid dyes and even Qdot® nanocrystals.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): E-Gel® Imager
Operating System: Windows 7, Windows XP (Service Pack 3)
Product Line: E-Gel™
Product Size: 1 each
Software Category: Validation and Verification Software
Software Type: Image Analysis
Shipping Condition: Room Temperature

Contents & storage

1 x Gel Quant Express software enabling USB Dongle.

Store at Room temperature.

Documents

Manuals & protocols

Recommended products