Ion Torrent™

Ion Sphere™ Quality Control Kit

Catalog number:  4468656

 Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ion Sphere Quality Control Kit furnishes a complete set of reagents for performing templated bead quality control using the Qubit 3.0 Fluorometer. This comprehensive kit contains calibration controls, buffers, and a probe mix for quality assessment of pre-enriched and post-enriched template-positive Ion Spheres. The powerful combination of the Ion Sphere Quality Control Kit and the Qubit 3.0 Fluorometer gives you greater experimental control and significant time and cost savings. By providing the confidence to generate high quality sequencing data with every run, this quality control process is ideal for those who are new to next-generation sequencing.

The Ion Sphere Quality Control Kit contains reagents for 20 quality control measurements. This kit is compatible with Qubit 2.0 and 3.0 fluorometers.

The Ion Sphere Quality Control Kit Gives You:
• Higher quality sequencing results through robust templated bead quality assessment
• A significantly quicker and more economical method for evaluating templated bead quality
• An increased level of experimental control and confidence

Rapidly Increase Your Proficiency in Semiconductor Sequencing
Quality control is essential for experimental success with new technologies. The Sphere Quality Control kit combined with the Qubit 3.0 Fluorometer offers a low cost, rapid, easy-to-use quality assessment solution, accelerating your proficiency in semiconductor sequencing. Don’t suffer by spending hours troubleshooting with “black-box" sequencers. Get high quality sequencing data the first time.

Enjoy Higher Confidence in Your Results
The Ion Sphere Quality Control Kit with the Qubit 3.0 Fluorometer brings greater experimental control to your science. For FFPE, LCM, or single cell applications where input material may be compromised, this robust assay determines sample quality prior to enrichment and sequencing, saving you time and money.

Benefit from a Quality Control Kit Designed for the Highly Accurate Qubit 3.0 Fluorometer
With state-of-the-art optical components, the capability for sophisticated data analysis, and an intuitive user interface, the Qubit 3.0 Fluorometer platform provides highly accurate and specific measurements not only of templated Ion Sphere Particles, but also of DNA, RNA, or protein. Through the use of dye-based quantitation, the Qubit 3.0 Fluorometer enhances your productivity by letting you avoid costly repetition of experiments.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibration Type: Sequencing - Templated Bead QC
Emission Class: Visible
Excitation Class: Visible
For Use With (Equipment): Ion PGM™ System, Ion S5™ System, Ion S5™ XL System, Qubit® Fluorometer
Product Line: Ion Sphere™, Ion Torrent™
Product Size: 1 kit
Quantity: 2 tube(s)
Volume (Metric): 800 µl
Workflow Step: Bead Deposition

Contents & storage

Store components at -20° C

Documents

Manuals & protocols