Applied Biosystems™

FLoid® Printer, Paper Cartridge, Ink/Paper

Catalog number:  4472351

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a complete printer package for the FLoid® Cell Imaging Station with all you need to immediately print your images and share or paste into your lab notebook. It includes a small, high quality printer, paper cartridge, ink, and paper.

This package provides you with:

Quality prints of your fluorescent cell images acquired on the FLoid® Cell Imaging Station

• Water resistant photos that last up to 100 years and are printed in just 47 seconds
• Paper with adhesive backing so photos can be pasted directly into your lab notebook

Increase the Productivity of Your FLoid® Cell Imaging Station
This compact photo printer is optimal for the FLoid® cell imaging station and prints photo lab quality images of your fluorescently labeled samples in under a minute. Print beautiful, borderless credit card sized photos in vibrant colors. Sticker-based backing allows you to paste directly into your lab notebook. It is an ideal printer for the FLoid® Cell Imaging Station that allows you to acquire three color fluorescent images in a minute. As the FLoid® Cell Imaging Station can sit right on your lab bench, not in some distant dark room, you can image and document your results wherever you want.

The prints will be water-resistant and can last up to 100 years! You'll enjoy photo lab quality and have fun all at the same time. Preview your images on the 2.5" Tilt-up LCD and easily turn your favorite pictures into an art-inspired collage with the image Shuffle feature.

Specifications

For Use With (Equipment): Floid® Cell Imaging System
Product Line: Floid®
Product Size: 1 kit
Shipping Condition: Room Temperature
Type: Printer

Contents & storage

Compact photo printer with credit card size paper cartridge, color ink and paper.

Store dry at room temperature.

Documents

Manuals & protocols