FLoid® Printer Ink/Paper

Catalog number: 4472389

Applied Biosystems™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Replacement color and ink paper set for the FLoid® Cell Imaging Station-compatible printer.

• All-in-one color ink cartridge and labels compatible with the FLoid® printer
• Color ink and label set produces credit card size label
• Adhesive back can be attached to your lab notebook
• Includes 18 sheets of credit card size dye sub labels
• Includes color ink ribbon for 18 prints


Print Beautiful, Borderless Credit Card Sized Photos in Vibrant Colors
Replacement color and ink paper set for the compact photo printer is optimal for the FLoid® cell imaging station (SKU # 44472351), which prints photo lab quality images of your fluorescently labeled samples in under a minute. The sticker-based backing allows you to paste directly into your lab notebook.

The prints will be water-resistant and can last up to 100 years! You'll enjoy photo lab quality and have fun all at the same time.

Specifications

For Use With (Equipment): Floid® Cell Imaging System
Product Line: Floid®
Product Size: 1 pack
Shipping Condition: Room Temperature
Type: Printer

Contents & storage

Color ink and paper set. 18 prints per pack.

Store dry at room temperature.

Documents

Manuals & protocols

Recommended products