Applied Biosystems™

Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems

Catalog number:  4472690

 Related applications: RNA Sequencing | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems is a reagent component of the AB Library Builder™ System. This kit, along with separately purchased Agencourt® AMPure® Beads, allows for the creation of up to 13 RNA whole transcriptome libraries on the Library Builder™.
• Automation – reduce hands on time by 50% or more depending on the number of libraries
• Scalable – prepare up to 13 libraries per run, up to 26 libraries per day
• Integrated – compatible with the SOLiD® 4 System and 5500 Series Genetic Analysis Systems

Labor Saving Automation
The Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems when used with the AB Library Builder™ System automates the most labor-intensive parts of RNA whole transcriptome library preparation, helping to reduce hands-on time by at least 50%. The Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems contains enough reagents and materials to process up to 13 samples at a time.

Integrated RNA Library Preparation
The Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems, along with the AB Library Builder™ System, is an integrated solution comprising a predefined software protocol, plug-and-play cartridge format reagents, and instrument. These kits are all optimized and validated for RNA purification and RNA whole transcriptome library preparation with the 5500 and SOLiD® 4.

To further maximize automation consider using RNA purification protocols with the iPrep™ PureLink® Total RNA Kit, compatible with the Library Builder™ System.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer, SOLiD™ 4 System
Library Type: Fragment Library
Workflow Step: Library Generation
Starting Material: RNA
Sequencing Type: Transcriptome Sequencing
Downstream Application: Next-Generation Sequencing
Final Product: cDNA Library
Product Line: Library Builder™
Product Size: 1 kit

Contents & storage

Ambient
Box A: 26 tips and tubes Box B: concentrator modules, tubes and buffers

Dry Ice
Reagent Box: 13 cartridges, Ligase buffer, RNAase III, primers and mastermix

Documents

Manuals & protocols

Recommended products