Applied Biosystems™

SYBR™ Select Master Mix for CFX

Catalog number:  4472942

 Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of SYBR Select Master Mix for CFX and added additional capabilities for your gene expression analysis.

**********************
SYBR® Select Master Mix for CFX is a superior and cost-effective solution for your real-time PCR applications.

• Specific—minimize primer-dimer and non-specific amplification
• Reproducible and sensitive—consistent amplification across a wide dynamic range
• Bright—contains SYBR® GreenER™ dye for maximum brightness
• Carry-over contamination control—contains heat-labile UDG
• For use with the Bio-Rad CFX96™ Touch or CFX384™ Touch systems
• Standard and fast cycling parameters

Formulated for Maximum Specificity
SYBR® Select Master Mix for CFX contains all the components needed for your real-time PCR reactions, except the template and primers, in a convenient 2X concentration premix. The master mix includes AmpliTaq® DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity.

Obtain Reproducible Results Across a Wide Dynamic Range
SYBR® Select Master Mix for CFX is specially formulated to provide robust results from 100 ng to 0.1 pg cDNA per reaction (Figure 1) and can reliably detect a single copy of genomic DNA (Figure 2).

Contains SYBR® GreenER™ Dye and Heat-labile UDG
• SYBR® GreenER™ dye is less inhibitory to PCR than SYBR® Green I dye, resulting in brighter signals
• Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control

View performance data.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: SYBR
For Use With (Equipment): Bio-Rad CFX Connect™, Bio-Rad CFX384 Touch™, Bio-Rad CFX96 Touch™
Reaction Speed: Fast, Standard
PCR Method: qPCR
Sample Type (General): DNA
Form: Liquid
Format: Vial(s)
Hot Start: Built-In Hot Start
Label or Dye: SYBR® GreenER™
Passive Reference Dye: ROX (Pre-mixed)
Polymerase: AmpliTaq® DNA Polymerase
Primer-Probe Compatibility: Unlabeled Oligos
Product Line: SYBR®
Product Size: 1 x 5 mL
Purity or Quality Grade: UP (Ultra Pure)
Sample Type (Specific): DNA (Genomic), cDNA
Shipping Condition: Wet Ice

Contents & storage

The 2x mix contains SYBR® GreenER™ dye, AmpliTaq® DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, and optimized buffer components. Contains 1 X 5 mL bottle, sufficient for 500 reactions at 20 µL. Store at 2-8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Documents

Manuals & protocols

Recommended products