E-Gel® Imager White-Light Conversion Screen

Catalog number: 4473061

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E-Gel® Imager White-Light Conversion Screen allows the visualization of protein gels on the E-Gel® Imager. This accessory can convert the blue light emitted by the E-Gel® Imager Blue-Light Base, as well as the UV light emitted by the E-Gel® Imager UV-Light Base, to white light to enable use of the powerful and intuitive E-Gel® Imager Software package for analysis and quantification of protein gels. The E-Gel® Imager White-Light Conversion Screen is compatible with protein gel stains such as SimplyBlue™ Safe , SilverQuest™ Silver, and Coomassie blue stains.

• A must-have upgrade to the E-Gel® Imager
• Ideal for the analysis of protein and silver-stained gels
• An affordable alternative to expensive protein-gel visualization instruments

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): E-Gel® Imager
Product Line: E-Gel®
Product Size: 1 each
Type: Conversion Screen
Shipping Condition: Room Temperature

Contents & storage

1 E-Gel® Imager White-Light Conversion Screen
Store at room temperature

Documents

Manuals & protocols

Recommended products