NUCLEIC-CARD™ matrix, 4 spots

Catalog number: 4473977

 Related applications: DNA Extraction | Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NUCLEIC-CARD™ is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature. The NUCLEIC-CARD™ matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD™ collection devices are available in one-, two-, and four-spot formats. They are manufactured by Copan, a market leader in the area of sample collection tools, and distributed by Thermo Fisher Scientific.

Key Features:

• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems® direct PCR amplification kits
• Certified free of DNase, Rnase, and amplifiable human DNA
• Prevents growth of microorganisms, enabling long-term room temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity

For Forensic or Paternity Use Only.

NUCLEIC-CARD™ is a trademark of Copan Flock Technologies.
For Forensic or Paternity Use Only.

Specifications

For Use With (Equipment): BSD Robotics Punching System, Harris Manual Punching System
Format: Card
Product Line: Applied Biosystems™, NUCLEIC-CARD™
Product Size: 100 cards
Shipping Condition: Room Temperature
Type: Sample Collection Accessory, Sample Storage Accessory

Contents & storage

100 NUCLEIC-CARD™ 4 Spots
Store at room temperature

Documents

Manuals & protocols

Recommended products