Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.

TaqMan® OpenArray® Human Kinome Panel, QuantStudio™ 12K Flex

Catalog number: 4475388

Applied Biosystems™  Related applications: Digital PCR | Pharma & Biopharma | Real Time PCR (qPCR) | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The TaqMan® OpenArray® Human Kinome Panel is an efficient, easy-to-use OpenArray® plate for the characterization of genes encoding human kinases. This panel targets 772 well-defined genes, plus 56 endogenous control genes. The format of the OpenArray® plate allows for 1–3 replicates to be run in parallel per plate.

The TaqMan® OpenArray® Human Kinome Panel is intended to capture the full set of protein kinases in the human genome. Genes were chosen based on the primary literature and public databases. In addition, there are 56 endogenous control genes against which the assays can be normalized. All assays in this panel have been bench-validated in our laboratories, so you can be sure that each assay is fully optimized.

The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.

This panel is intended for use with the QuantStudio™ 12K Flex Real-time PCR system which can cycle and image 4 pates simultaneously, providing high-throughput results with ease.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: OpenArray® Plate
Label or Dye: FAM™
Product Line: OpenArray®, TaqMan™
Product Size: 1 each
Reaction Speed: Standard
Research Category: Other Research
Species: Human
Target Organism Class: Human

Contents & storage

Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C

Documents

Manuals & protocols