Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Applied Biosystems™

TaqMan™ OpenArray™ Human Stem Cell Panel, QuantStudio™ 12K Flex

Catalog number:  4475390

 Related applications: Digital PCR | Real Time PCR (qPCR) | Stem Cell Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The TaqMan® OpenArray® Human Stem Cell Panel is an efficient, easy-to-use OpenArray® plate for the characterization of undifferentiated stem cells or their differentiated derivatives. This gene signature panel targets 609 well-defined genes validated as markers for pluripotency, plus 22 endogenous control genes. The format of the OpenArray® plate allows for 1–4 replicates to be run in parallel per plate.

The TaqMan® OpenArray® Human Stem Cell Panel contains a well-defined set of validated gene expression markers to characterize human embryonic stem (ES) cell identity and assess phenotypic variations between embryonic stem cell isolates. The gene content of this OpenArray® plate resulted directly from the work of the I.S.C.I consortium and characterization of human embryonic stem cell lines by the International Stem Cell Initiative.

The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.

The Right Targets
This OpenArray® plate uses the proprietary Applied Biosystems® OpenArray® technology to simultaneously analyze gene expression levels by quantitative PCR for 609 test genes and 22 endogenous controls.

Genes were included based on the following criteria:

• Expression in human embryonal carcinoma cells (hEC), primordial germ cells, or human ES (hES) cells
• Known role in maintenance of pluripotency in hES cells
• Identification in microplate-based experiments as being correlated with the stem cell state
• Expression known to be characteristic of specific differentiation lineages in the post implantation embryo and mature tissues

This panel is intended for use with the QuantStudio™ 12K Flex Real-time PCR system which can cycle and image 4 pates simultaneously, providing high-throughput results with ease.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: OpenArray™ Plate
Label or Dye: FAM™
Product Line: OpenArray®, TaqMan™
Product Size: 1 each
Reaction Speed: Standard
Research Category: Stem Cell Research
Species: Human
Target Organism Class: Human

Contents & storage

Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C

Documents

Manuals & protocols