Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.

TaqMan™ OpenArray™ Human Cancer Panel, QuantStudio™ 12K Flex

Catalog number:  4475391

Applied Biosystems™  Related applications: Digital PCR | Oncology & Genetic Disease Research | Pharma & Biopharma | Real Time PCR (qPCR) | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The TaqMan® OpenArray® Human Cancer Panel is an efficient, easy-to-use OpenArray® plate for the characterization of undifferentiated cancers or their differentiated derivatives. This gene signature panel targets 624 well-defined genes validated as markers for pluripotency, plus 24 endogenous control genes. The format of the OpenArray® plate allows for 1–4 replicates to be run in parallel per plate.

The TaqMan® OpenArray® Human Cancer Panel targets genes related to DNA repair, angiogenesis, cell adhesion, and ECM, as well as genes involved in the cell cycle and apoptosis, and many of the genes encoding kinases and transcription factors that have been found to be differentially expressed in early cancer and metastatic disease. In addition, 24 endogenous control genes are included against which the assays can be normalized. All assays in this panel were developed using a high level of bioinformatics and wet-lab stringency.

The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.

This panel is intended for use with the QuantStudio™ 12K Flex Real-time PCR system which can cycle and image 4 pates simultaneously, providing high-throughput results with ease.
For Research Use Only. Not for use in diagnostic procedures.


Format: OpenArray® Plate
Label or Dye: FAM™
Product Line: OpenArray®, TaqMan™
Product Size: 1 each
Reaction Speed: Standard
Research Category: Oncology
Species: Human
Target Organism Class: Human

Contents & storage

Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C


Manuals & protocols