Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.

TaqMan® OpenArray® Mouse Inflammation Panel, QuantStudio™ 12K Flex

Catalog number: 4475393

Applied Biosystems™  Related applications: Digital PCR | Pharma & Biopharma | Real Time PCR (qPCR) | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The TaqMan® OpenArray® Mouse Inflammation Panel is an efficient, easy-to-use OpenArray® plate for quantitative gene expression analysis of mouse inflammation genes important in drug discovery.

The panel covers 632 genes that have been studied as targets for a range of inflammatory diseases, plus 16 endogenous control genes. The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.

This panel is intended for use with the QuantStudio™ 12K Flex Real-time PCR system which can cycle and image 4 pates simultaneously, providing high-throughput results with ease.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: OpenArray® Plate
Label or Dye: FAM™
Product Line: Applied Biosystems™, OpenArray®, TaqMan™
Product Size: 1 each
Reaction Speed: Standard
Research Category: Inflammation
Species: Mouse
Target Organism Class: Mouse

Contents & storage

Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C

Documents

Manuals & protocols