Invitrogen™

DynaLight™ Trigger Solution

Catalog number:  4475403

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Achieve faster ELISA results with DynaLight™ Trigger Solution when used in combination with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized to achieve faster results in solution-based assays.

Features of DynaLight™ Substrate with RapidGlow™ Enhancer:

• Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
• Sensitive attomole detection of purified alkaline phosphatase enzyme
• Optimized formulation for both microplate and bead-based assays
• Substrate stability of over 1 year at 4°C
• Compatible with automated platforms with trigger solution/positions already in place

The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Specifications

Assay Type: Immunoassay
Compatible Cells: Mammalian Cells
Detection Method: Chemiluminescent
Detection Range or Limit: pg/ml to µg/ml
For Use With (Equipment): Luminometer (Tube-Based)
Form: Liquid
Format: Bottle(s)
High Throughput Compatibility: High Throughput-Compatible
Product Line: DynaLight™
Product Size: 100 mL
Reaction Time: 31 sec
Readout: End Point
Sample Type (Specific): Cell Cultures, Serum, Whole Blood
Sensitivity: 1 pg⁄ml
Substrate: DynaLight™ Substrate
Substrate Properties: Chemical Substrate
Substrate Type: AP (Alkaline Phosphatase) Substrate
Target Enzyme: Alkaline Phosphatase
Shipping Condition: Room Temperature

Contents & storage

1 clear bottle. Store at room temperature.

Documents

Manuals & protocols

Recommended products