Need help deciding what product to use with the Ion PGM™ System? Check out our helpful Applications Decision Tree for the Ion PGM™ System.
Ion Torrent™

Ion Xpress™ RNA-Seq Barcode 1-16 Kit

Catalog number:  4475485

 Related applications: Epigenetic Sequencing | Ion Torrent™ Next-Generation Sequencing | RNA Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The new Ion Xpress™RNA-Seq Barcode 1-16 Kit provides a set of 16 unique barcode primers for sequencing RNA on the Ion Torrent PGM™semiconductor sequencer. When used in combination with the Ion Total RNA-Seq Kit v2, the Ion Xpress™RNA-Seq Barcode 1-16 Kit enables users to pool up to 16 fragment libraries prior to template preparation and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for small and whole transcriptome RNA.

Features and Benefits:
• Efficient multiplexing-enables multiplexing of up to 16 fragment libraries on a single sequencing chip
• Both sequence- and flow-optimized for equal representation of all barcodes in a pool
• Multiplexing decreases the cost and the handling requirements of template preparation and sequencing
• Robust error correction built in by design for added confidence in sample identification

Efficient Multiplexing with Barcode Primers Enables Higher Throughput Sample Sequencing
Multiplexing with barcoded libraries leads to more cost-effective runs by permitting up to 16 samples per run, significantly decreasing the cost and handling requirements of template preparation and sequencing.

Sequence- and Flow-Optimized for Increased Performance and Efficiency
Barcode primers contain sequences that are optimized to provide equal representation of all barcodes in a pool, require a minimum number of flows to interrogate, and necessitate only 20 bases of barcode and adaptor sequencing, which results in more economical multiplexed sequencing runs.

Robust Error Correction Built In By Design
Built-in error correction significantly minimizes the possibility of incorrect sample assignment.

The Ion Xpress™ RNA-Seq Barcode 01-16 Kit is designed to create 10 multiplexed libraries per barcode or a total of 160 multiplexed libraries.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion PGM™ System
Workflow Step: Library Barcoding
Product Size: 160 reactions
Shipping Condition: Dry Ice

Contents & storage

16 tubes, 12 µL each Ion Xpress™ RNA BC 01 - BC 16 - Store at 20 °C

1 tube, 192 µL Ion Xpress™ RNA 3' Barcode Primer - Store at 20 °C

Documents

Manuals & protocols