Ion Plus and Ion Xpress™ Plus Library Protocol Card for AB Library Builder™ System

Catalog number: 4477687

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ion Plus and Ion Xpress™ Plus Library Protocol Card for AB Library Builder™ System is required for running the Ion Xpress™ Plus Fragment Library Kit for AB Library Builder™ System and the Ion Plus Fragment Library Kit for AB Library Builder™ System. This protocol card contains the firmware needed to easily automate the creation of up to 13 Ion Plus or Ion Xpress™ Plus DNA fragment libraries containing standard, paired-end, or barcoded adapters upstream of semiconductor sequencing on either the Ion PGM™ or Ion Proton™ platforms.

The Ion Plus and Ion Xpress™ Plus Library Protocol Card for AB Library Builder™ System:
• Enables Ion Xpress™ Plus and Ion Plus fragment library construction on AB Library Builder™ System
• Eliminates the need for instrument programming prior to use

The protocol card also offers:
• Convenience of having instrument software for automating DNA fragment library preparation on the AB Library Builder™ System on an easy-to-install firmware card
• Simplicity of running multiple protocols without the need for instrument programming or time consuming training

A Single-Day Workflow: The Next Stage in a Sequencing Revolution
With the ability to prepare up to 13 libraries in a little over 2 hours, the Ion Plus and Ion Xpress™ Plus fragment library kits for the AB Library Builder™ System represent a step towards automating the Ion sequencing workflow, making comprehensive analyses of your sample(s) of interest more convenient than ever before. This protocol card makes implementing these automated workflows simple and easy, for a transition from manual to automated library construction that is rapid and painless.

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): AB Library Builder™
Product Line: Applied Biosystems™, Ion Torrent™, Ion Xpress™, Library Builder™
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Card

Contents & storage

1 firmware card

Store at room temperature.

Documents

Manuals & protocols