VeriFiler™ Direct PCR Amplification Kit

Catalog number:  4479567

Applied Biosystems™  Related applications: Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The VeriFiler™ Direct PCR Amplification Kit is a multiplex assay designed to amplify 9 unique STR loci plus the sex determining marker amelogenin. The kit’s loci configuration is optimized for use with the Identifiler® series of kits to enable increased discriminatory power when needed for your difficult paternity cases. In addition to its ability to amplify from gDNA extracts, the kit is validated for direct amplification from common single-source samples. Direct amplification eliminates the need for gDNA extraction and quantification, minimizing hands on time and labor requirements. The VeriFiler™ Direct kit helps you get more information using your existing instrumentation and workflow.

The VeriFiler™ Direct PCR Amplification Kit:
• Targets these 9 STR loci: TH01, D2S1338, D19S433, D6S1043, D1S1656, D2S441, D10S1248, D12S391, D22S1045
• Employs a direct amplification workflow from these common sample types used in paternity testing: buccal swabs, FTA card punches, gDNA extracts
• Works with your existing Applied Biosystems® CE instrumentation and data analysis software packages: Veriti® and 9700 thermal cyclers, 3130 and 3500 genetic analyzers, GeneMapper® ID and GeneMapper® ID-X software

When run with Identifiler® Series products, the VeriFiler™ Direct kit:
• Yields 6 additional highly discriminatory STR loci
• Amplifies 3 overlapping STR loci to confirm inter-assay genotyping consistency
• Further decreases probability of identity by about 8 orders of magnitude and increases the probability of paternity exclusion

Important Notice

The purchase of this product conveys to the buyer the non-transferable license to use the purchased amount of the product only for paternity testing conducted by the buyer. This license does not include, as ruled by a U.S. Court, a license to use this product in chimerism determination or analysis, molar specimens classification, cell line authentication, determination of fetal sex, cancer analysis, genetic research, non-casework-related forensic applications (such as general research in forensics or teaching and training persons not employed in a forensic laboratory), identifying or determining maternal cell contamination, or sample tracking. Further rights can be obtained by contacting Promega Corp., 2800 Woods Hollow Rd., Madison, WI 53711 USA.
For Paternity Use Only. Not for use in Research or Clinical Diagnostic applications.

Specifications

Amplicon Size: <300 bp
For Use With (Equipment): 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, GeneAmp™ 9700, Veriti® Thermal Cycler
Label or Dye: FAM™, LIZ™, NED™, PET™, VIC®
Matrix and Size Standards: Matrix Standard: DS-33 Matrix Standard Kit (Dye Set G5), Size Standard: GeneScan&trade 600 LIZ® Size Standard v2.0
Polymerase: AmpliTaq Gold® DNA Polymerase
Product Line: VeriFiler™
Product Size: 200 tests
Reaction Format: SuperMix or Master Mix
Sample Volume: 25 µL⁄reaction
Technique: STR (Short Tandem Repeat) Analysis

Contents & storage

Store at -15 to -25 °C on receipt and 2 to 8 °C after initial use.

Documents

Manuals & protocols