Applied Biosystems™

Quantifiler™ HP DNA Quantification Kit

Catalog number:  4482911

 Related applications: Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Quantifiler® HP (Human Plus) DNA Quantification Kit is a robust kit that enables forensic laboratories to simultaneously obtain a quantitative and qualitative assessment of total human DNA in a single, highly sensitive real-time PCR reaction. This guides selection of the optimal short tandem repeat (STR) analysis chemistry (autosomal or miniSTR) and streamlines the workflow while increasing downstream analysis success rates. The kit includes enhanced quantity and quality indicators that help deliver accurate, sensitive, and specific results that work together with the HID Real Time PCR Analysis Software v1.2 to enable informative and integrated workflow decisions. The Quantifiler® HP kit and the HID v1.2 software are designed for use with the Applied Biosystems® 7500 Real-Time PCR System.

Highly Sensitive
• <1 pg/µL limit of detection. Quantifies DNA concentrations from 0.005 ng/uL to >50 ng/uL, enabling the use of a broad range of concentrations, with flexible protocols for higher concentration samples.

Highly Specific
• Detects human and higher primate DNA

Highly Informative
• Quantifies DNA from a wide variety of sources, including buccal swabs, blood, semen, tissue, and other human DNA sources, making this kit suitable for casework sample screening, databasing, and paternity labs
• Guides selection of the optimal AmpFLSTR® PCR Amplification Kit (autosomal or miniSTR)
• Quality Index helps improve prediction of STR amplification performance and estimation of DNA quantity in degraded samples, especially in the presence of inhibitors.
• Enhanced inhibition detects the presence of PCR inhibitors (suggesting additional purification of extract and/or whether a mini STR kit like the MiniFiler™, NGM SElect™, or GlobalFiler™ kit may be necessary).

Highly Reliable
• Uses gold-standard TaqMan® real-time PCR technology already widely adopted in forensic laboratories. Real-time PCR has the widest dynamic range of any DNA detection technology allowing for more accurate quantification of low level samples.

Highly Efficient
• The quantification system minimizes sample consumption and streamlines workflow, with results in ~1 hour.
• Optimized and validated as part of a complete DNA testing system with the Applied Biosystems® 7500 Real-Time PCR System, AmpFLSTR® PCR Amplification kits and HID Real Time PCR Analysis Software v 1.2 to enable high quality STR profiling results.

A Highly Informative Workflow Tool
The Quantifiler® HP DNA Quantification Kit has been optimized and validated as part of a complete DNA testing system with the AmpFLSTR® PCR Amplification kits. The chemistry is compatible with commonly used extraction technologies for easy integration into current lab workflows.This enables the Quantifiler® HP kit to function as a critical component of an improved and integrated workflow for more effective processing of forensic casework samples. Both the kit and the software are fully automatable with most liquid handling robotic platforms.

Comparison to Quantifiler® Human Kits
There are several specific differences between the Quantifiler® HP DNA Quantification Kit and the Quantifiler® Human DNA Quantification Kit (Cat. No. 4343895), including:
Target Loci: The Quantifiler® HP kit uses multiple-copy target loci for improved detection sensitivity. There are two human-specific target loci: Small Autosomal and Large Autosomal. Each consists of multiple copies dispersed on various autosomal chromosomes.
Optimized Chemistry: The assay chemistry has been optimized for more efficient multiplexing, faster PCR cycle
times (~1 hour), and better inhibitor tolerance. The overall performance improvements allow the Quantifiler® HP kit to better match the enhanced performance of newer STR kits that are designed to accommodate more challenging samples, like the Identifiler® Plus, NGM SElect™, or GlobalFiler™ PCR Amplification kits.
DNA Standard: The Quantifiler® HP DNA Standard is pooled, human genomic, smaller molecular weight DNA for improved stability, homogeneity, and correlation with forensic samples. There is a new five-point standard curve protocol that helps improve efficiency with more samples on a plate.
Packaging Features: To help laboratories control for and minimize the potential of reagent contamination, as well as to better accommodate automated platforms, the Quantifiler® HP DNA Quantification Kit has some key packaging features:
    --Four 100-reaction individual-use screw cap tubes (1 tube will process a 96 well plate)
    --The DNA standard is packaged in a separate box.
    --Both the reagent and DNA standard box are shipped together in an STR-style sleeve with new peel & stick reagent lot barcode labels. This allows customers to separate out the reagents easily for special storage upon arrival at the laboratory.
For Forensic or Paternity Use Only.

Specifications

Product Line: Quantifiler™
Product Size: 400 reactions
Target Gene: large autosomal (LA) human target, small autosomal (SA) human target

Contents & storage

Store at -5 to -30°C upon receipt, 2–8°C after initial use.

Documents

Manuals & protocols

Recommended products