Notice
: The Ion PGM™ IC 200 Kit will be discontinued in Q1 of 2017. The supported replacement kit, the Ion PGM™ Hi-Q™ View Chef Kit (A29902), is now available for order. The final build of the Ion PGM™ IC 200 Kit will be Q1 of 2017 and will have a 7–8 month shelf life when stocked. Based on in-stock availability, this will allow for migration to the Ion PGM™ Hi-Q™ View Chef Kit (A29902) until the end of 2017.
 

Ion PGM™ IC 200 Kit

Catalog number: 4484080

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ion PGM™ IC 200 Kit enables robust and reproducible template preparation, chip loading, and sequencing of 200 base-read libraries on the Ion Chef™ system for the Ion PGM™ system sequencing workflow. The plug & play, pre-packaged single-use reagent cartridges for template and chip preparation provide a fully automated workflow, processing dual samples per run for increased productivity. The reagent and sample barcoding features interact with the Ion Chef™ system to validate user-defined run plan and offer complete run traceability.

The Ion PGM™ IC 200 Kit offers:

• Fully automated and reproducible template preparation, chip loading, and sequencing of 200 base-read libraries
• Less than 15 minutes hands-on time
• Ready-to-use, disposable reagent cartridges
• Increased throughput with dual sample processing
• Multiplexing capability
• Increased robustness for quality sequencing results

The Ion PGM™ IC 200 Kit is an integral component of the Ion Torrent™ next-generation sequencing workflow, designed to simplify and streamline the overall workflow with minimal hands-on-time for users with any experience level. This easy-to-use kit delivers high-quality and reproducible templated products for semiconductor sequencing.

The Ion PGM™ IC 200 Kit contains sufficient reagents and consumables for 8 template preparation and sequencing runs, and should be used in conjunction with Ion 314™, Ion 316™, or Ion 318™ v2 barcoded chips for sample tracking purposes.

Notes:

• This kit supports 200 base-read libraries for a variety of applications.
• This kit is only compatible with the Ion Chef™ system and the Ion PGM™ system.
• Components of this kit have been optimized for use on the Ion Chef™ system and the Ion PGM™ system specifically; and should NOT be replaced or interchanged with other product kit components.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Ion Chef™ System, Ion PGM™ System
Library Type: 200 base-read libraries
Workflow Step: Chip Loading, Sequencing, Template Preparation
Sequencing Type: Genome & DNA Sequencing
Chip Compatibility: Ion 314™ Chip v2 BC, Ion 316™ Chip v2 BC, Ion 318™ Chip v2 BC
High Throughput Compatibility: Automated Protocols, High Throughput-Compatible, Multiplexing
Product Line: Ion Chef™
Product Size: 8 reactions

Contents & storage

The Ion PGM™ IC 200 Kit components include template reagents, enzyme mixes, nucleotides, spike-in sequencing controls, solutions & buffers, consumables, and supplies.

Documents

Manuals & protocols

Recommended products