Informational:This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
These TaqMan™ assay beads provide a simple, reliable, and rapid method for detecting the presence of a specific bacterial or viral target. The assay uses PCR to amplify a DNA target unique to the microorganism and TaqMan™ probe to detect and quantify the target, all in an an easy-to-use, lyophilized bead format. TaqMan™ assay beads deliver the fastest RT-PCR run times on the market (<40 minutes in Fast mode). Simply add 30 μL of purified DNA to the beads and run on Fast or standard instrumentation. These TaqMan™ assay beads provide microbial detection capabilities and high-confidence results to help accelerate your food and environmental testing needs. The assay design is based on US EPA published guidelines and validated by the EPA.
Features of TaqMan™ assay beads:
• Convenience—simply add your nucleic acid sample to the bead-containing assay tube and run; no pre-mixing or master mix preparation required • Consistency—fewer operator steps means reduced error and increased reproducibility across time, users, and laboratories • Improved data accuracy—an internal positive control is included to monitor your PCR • Fast chemistry—40-min run time with Fast cycling, compatible with standard or Fast modes
TaqMan™ assay beads are suitable for single or multiplex analysis. Because they are run using universal conditions, multiple bead-based assays can be analyzed in the same run. The robust assay design provides highly accurate, reproducible, and reliable results, with the enhanced stability of a lyophilized format. A passive internal reference based on proprietary ROX™ dye is included for precise data analysis.
This product is for environmental testing, quality control/quality assurance testing, food and agricultural testing, and research purposes only.
Specifications
Description
Includes Master Mix
Detection Method
Primer-probe
Form
Lyophilized
Reaction Speed
Fast or Standard
Species
All Species
Sufficient For
96 Reactions
Target Organism Class
Enterococcus spp.
For Use With (Application)
Food & Beverage
No. of Reactions
96 Reactions
Contents & Storage
• 12 8-tube strips containing assay beads (store at 2–8°C in the dark) • Optical 8-tube strips and 8-cap strips (store at room temperature)