QuantStudio™ 7 Flex Real-Time PCR System, array card, laptop

Catalog number: 4485700

Applied Biosystems™  Related applications: Digital PCR | Genotyping & Genomic Profiling | Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems® QuantStudio™ 7 Flex Real-Time PCR System advances your research further by enabling a broad range of real-time PCR-based applications through its multiplexing capabilities and interchangeable block formats. With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.

Features of the QuantStudio™ 7 Flex Real-Time PCR System include:

• Increased application versatility through the accomodation of 4 different block types
• Compatible with over 8 million TaqMan® assays for gene expression, genetic variation, gene regulation, or protein expression experiments
• The intelligent design of the QuantStudio™ platform combined with Applied Biosystems® instrument reliability and accuracy
• Easy-to-use software, responsive touch-screen, automation capabilities, and effortless block exchange without the need for any tools

Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

>> Download a 90-day trial of the QuantStudio™ 7 software

The QuantStudio™ 7 Flex system accommodates the interchange of a 96-well, 96-well Fast, 384-well, or TaqMan® array card block. The system is delivered with one block type (an array card block in this case); other block types may be purchased separately. In addition, a Twister® Robot Automation Accessory is available for this system to support high throughput workflows. For a more value-priced system that does not support TaqMan® array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio™ 6 Flex system. And for the ultimate in flexibility, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan® OpenArray® plates.

Accomplish More, Faster
Run hundreds of real-time PCR reactions effortlessly using TaqMan® Array 384-well microfluidic cards. And when integrated with the Twister® Robot Automation Accessory (available separately), the QuantStudio™ 7 Flex system maximizes your throughput capabilities.

High Application Versatility
Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, High Resolution Melt (HRM), and Protein Thermal Shift™ experiments.

Results You Can Trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex™ system, which features 6 decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.

Sensitive Data Analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Block Format: Interchangeable
For Use With (Equipment): QuantStudio™
Format: 384-well microfluidic card
Product Line: QuantStudio™
Product Size: 1 system

Documents

Manuals & protocols