Beginning August 15, 2014, this product will ship under ambient conditions. Full details can be found here.
Applied Biosystems™

TaqMan™ GAPDH Assay, JUN™ dye/QSY™ probe This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  4485712

 Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems® TaqMan® GAPDH Assay with JUN® dye-labeled QSY® probe provides a pre-formulated assay for quantitating human GAPDH. This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. The assay consists of a JUN® dye-labeled QSY® probe plus sequence-specific forward and reverse primers, and it can be used for multiplex or singleplex PCR reactions.

Benefits of this assay include:

• Eliminate assay design time by using pre-designed primers and pre-designed JUN® dye-labeled TaqMan® probe
• Minimize experimental optimization time
• Use in conjunction with FAM™ or VIC® dye-labeled predesigned gene expression assays

Save time in assay design, formulation, and testing by using the TaqMan® GAPDH Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.

The JUN® dye is optimized for use with the 4th filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments. Please note that master mix containing ROX™ dye cannot be used with this assay (with JUN® dye) due to an overlap of spectra. TaqMan® Multiplex Master Mix with MUSTANG PURPLE® dye is recommended.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Accession Number: NM_002046.3
Amplicon Size: 118
Format: Tube(s)
Gene Symbol: GAPDH
Includes: Primer-Probe Set(s)
Internal Probe Modification: QSY® Non-Fluorescent Quencher (3')
Label or Dye: JUN™
Product Line: TaqMan™
Product Size: 250 reactions
Species: Human
Target Gene: Glyceraldehyde-3-phosphate dehydrogenase
Green Features: Sustainable packaging

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products