BigDye® Terminator v3.1 Sequencing Standard-CG, 3730 Series

Catalog number: 4486194

Applied Biosystems™  Related applications: Diagnostic Development | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BigDye® Terminator v3.1 Sequencing Standard-CG is a General Purpose Reagent that consists of four tubes, each of which contains enough hsp69 DNA of a known sequence prepared with BigDye® Terminator v3.1 to perform one spectral calibration/performance check and/or one control sequencing run on an Applied Biosystems® 3730 Series genetic analyzer. The DNA contained in the preparation has been lyophilized to maximize stability. This standard is for use with capillary-electrophoresis–based genetic analyzers and is not validated for use with other systems or applications. These reagents are For Laboratory Use.
For Laboratory Use

Specifications

Label or Dye: Dye Set Z
Product Size: 4 runs
For Use With (Equipment): 3730 DNA Analyzer

Documents

Manuals & protocols