GABA Transporter 3 Antibody

Catalog number: 480018

Invitrogen™  Related applications: Cellular Imaging | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

catalyzes sodium and chloride dependent transport of gamma-aminobutyric acid (GABA) to terminate GABA mediated synaptic activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Rat
Validated Application: Western Blot, Immunohistochemistry
Host Species: Rabbit
SubClass (Isotype): IgG1
Clonality: Polyclonal
Clone ID: pAB
Antibody Fragment: Whole Antibody
Target Gene: SLC6A11
Gene Symbol: SLC6A11
Label or Dye: Unconjugated
Purification: Affinity Purified
Form: Frozen
Product Size: 100 µL
Volume (Metric): 100 µl
Shipping Condition: Wet Ice

Contents & storage

2-8°C for up to 1 month, -20°C for long term storage. Avoid repeated freezing and thawing.

Documents

Manuals & protocols

Recommended products