DynaMag™-PCR Magnet

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The MAGnify™ ChIP system utilizes a novel magnet that is compatible with 0.2mL PCR strip tubes. This magnet is critical for enabling the use of reduced starting cell numbers and volumes for immunoprecipitation and washing steps. This novel magnetic separator allows you to achieve results quickly and easily while offering the freedom to simultaneously perform multiple ChIP assays in PCR tubes with a multi-channel pipettor.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 1 each
Shipping Condition: Room Temperature
For Use With (Equipment): DynaMag™

Contents & storage

Store magnet at room temperature.

Documents

Manuals & protocols

Recommended products